Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human PD-L1/B7-H1 Alexa Fluor™ 488-conjugated Antibody, R&D Systems™



Mouse Monoclonal Antibody has been used in 2 publications
$245.00 - $526.00
Specifications
| Antigen | PD-L1/B7-H1 |
|---|---|
| Clone | 130021 |
| Dilution | Flow Cytometry 5 uL/10^6 cells |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
FAB1561G025
![]() |
R&D Systems
FAB1561G025 |
25 Tests |
Each for $245.00
|
|
|||||
FAB1561G100
![]() ![]() |
R&D Systems
FAB1561G100 |
100 Tests |
Each for $526.00
|
|
|||||
Description
PD-L1 Monoclonal specifically detects PD-L1 in Human samples. It is validated for Flow Cytometry.Specifications
| PD-L1/B7-H1 | |
| Flow Cytometry 5 uL/10^6 cells | |
| Monoclonal | |
| Purified | |
| RUO | |
| Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide | |
| Avelumab, B7-H, B7H1, B7-H1, B7H1PDCD1L1, CD274, CD274 antigenMGC142294, CD274 molecule, PDCD1L1, PDCD1LG1, PDCD1LG1MGC142296, PDL1, PD-L1, PD-L1B7 homolog 1, PDL1PDCD1 ligand 1, programmed cell death 1 ligand 1, Programmed death ligand 1 | |
| CD274 | |
| IgG1 | |
| Protein A or G purified from hybridoma culture supernatant | |
| Detects human B7-H1/PD-L1 in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant human (rh) B7-1, -2, -H2, -H3, -H3b, -H4, rhPD-L2, recombinant mouse B7-H1, recombinant rat (rr) B7-1, or rrB7-2 is observed. |
| 130021 | |
| Flow Cytometry | |
| Alexa Fluor 488 | |
| Mouse | |
| Human | |
| Q9NZQ7 | |
| 29126 | |
| Mouse myeloma cell line NS0-derived recombinant human B7-H1/PD-L1 Phe19-Thr239 Accession # Q9NZQ7 | |
| Primary | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title