Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human PD-L1/B7-H1 APC-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. FAB1561A100
Change view
Click to view available options
Quantity:
100 Tests
25 Tests
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FAB1561A100 100 Tests
FAB1561A025 25 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. FAB1561A100 Supplier R&D Systems Supplier No. FAB1561A100
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

PD-L1 Monoclonal specifically detects PD-L1 in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen PD-L1/B7-H1
Applications Flow Cytometry
Classification Monoclonal
Clone 130021
Conjugate APC
Dilution Flow Cytometry 10 uL/10^6 cells
Formulation Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide
Gene Accession No. Q9NZQ7
Gene Alias Avelumab, B7-H, B7H1, B7-H1, B7H1PDCD1L1, CD274, CD274 antigenMGC142294, CD274 molecule, PDCD1L1, PDCD1LG1, PDCD1LG1MGC142296, PDL1, PD-L1, PD-L1B7 homolog 1, PDL1PDCD1 ligand 1, programmed cell death 1 ligand 1, Programmed death ligand 1
Gene Symbols CD274
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human B7-H1/PD-L1 Phe19-Thr239 Accession # Q9NZQ7
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 Tests
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 29126
Test Specificity Detects human B7-H1/PD-L1 in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant human (rh) B7-1, -2, -H2, -H3, -H3b, -H4, rhPD-L2, recombinant mouse B7-H1, recombinant rat (rr) B7-1, or rrB7-2 is observed.
Target Species Human
Content And Storage Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.