Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human PD-L2/B7-DC Alexa Fluor™ 594-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AB12241T100
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AB12241T100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AB12241T100 Supplier R&D Systems Supplier No. FAB12241T100UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

PD-L2/B7-DC/PDCD1LG2 Monoclonal specifically detects PD-L2/B7-DC/PDCD1LG2 in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen PD-L2/B7-DC/PDCD1LG2
Applications Flow Cytometry
Classification Monoclonal
Clone 988708
Conjugate Alexa Fluor 594
Dilution Flow Cytometry 0.25-1μg/106 cells
Formulation Supplied 0.2Mg/mL in a saline solution containing BSA and Sodium Azide.
Gene Accession No. Q9BQ51
Gene Alias B7-DC, bA574F11.2, Btdc, Butyrophilin B7-DC, Butyrophilin-like Protein, CD273, CD273 antigen, CD273PD-1 ligand 2, MGC142240, PD-1-ligand 2, PDCD1L2MGC142238, PDCD1LG2, PDL2, PDL2B7DC, PD-L2PDCD1 ligand 2, programmed cell death 1 ligand 2, Programmed death ligand 2
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human PD-L2/B7-DC Met1-Pro219 Accession # Q9BQ51
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) 80380
Test Specificity Detects human PD-L2/B7-DC in direct ELISAs.
Target Species Human
Content And Storage Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied.
Form Supernatant
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.