Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human PD-L2/B7-DC Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 3 publications
$214.00 - $596.50
Specifications
Antigen | PD-L2/B7-DC/PDCD1LG2 |
---|---|
Concentration | LYOPH |
Dilution | Western Blot 2 ug/mL, Immunohistochemistry 5-15 ug/mL, Blockade of Receptor-ligand Interaction |
Applications | Western Blot, Immunohistochemistry, Blockade of Receptor-ligand Interaction |
Classification | Polyclonal |
Description
PD-L2/B7-DC/PDCD1LG2 Polyclonal specifically detects PD-L2/B7-DC/PDCD1LG2 in Human samples. It is validated for Western Blot, Immunohistochemistry, Blockade of Receptor-ligand Interaction.Specifications
PD-L2/B7-DC/PDCD1LG2 | |
Western Blot 2 ug/mL, Immunohistochemistry 5-15 ug/mL, Blockade of Receptor-ligand Interaction | |
Polyclonal | |
Goat | |
Human | |
B7-DC, bA574F11.2, Btdc, Butyrophilin B7-DC, Butyrophilin-like Protein, CD273, CD273 antigen, CD273PD-1 ligand 2, MGC142240, PD-1-ligand 2, PDCD1L2MGC142238, PDCD1LG2, PDL2, PDL2B7DC, PD-L2PDCD1 ligand 2, programmed cell death 1 ligand 2, Programmed death ligand 2 | |
PDCD1LG2 | |
IgG | |
Affinity Purified | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
LYOPH | |
Western Blot, Immunohistochemistry, Blockade of Receptor-ligand Interaction | |
Unconjugated | |
RUO | |
Q9BQ51 | |
80380 | |
Mouse myeloma cell line NS0-derived recombinant human PD-L2/B7-DC Leu20-Pro219 Accession # Q9BQ51 | |
Primary | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Detects human PD-L2/B7-DC in direct ELISAs and Western blots. In direct ELISAs, approximately 50% cross-reactivity with recombinant mouse PD-L2 is observed and less than 1% cross-reactivity with recombinant human (rh) B7-1, rhB7-2, rhB7-H1, rhB7-H2, rhB7-H3, rhB7-H3b and recombinant rat B7-1 is observed. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title