Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human PDCD4 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems MAB7019SP
This item is not returnable.
View return policy
Description
PDCD4 Monoclonal specifically detects PDCD4 in Human samples. It is validated for Immunocytochemistry.Specifications
PDCD4 | |
Monoclonal | |
LYOPH | |
Immunocytochemistry 8-25 ug/mL | |
Q53EL6 | |
PDCD4 | |
E. coli-derived recombinant human PDCD4 Lys212-His358 Accession # Q53EL6 | |
25 μg | |
Primary | |
Detects human PDCD4 in direct ELISAs. | |
Human | |
Purified |
Immunocytochemistry | |
808508 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
H731, H731nuclear antigen H731, MGC33046, MGC33047, Neoplastic transformation inhibitor protein, Nuclear antigen H731-like, programmed cell death 4 (neoplastic transformation inhibitor), programmed cell death protein 4, Protein 197/15a | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
27250 | |
Sterile PBS to a final concentration of 0.5 mg/mL. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction