Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human PDGF-AA Antibody, R&D Systems™

Mouse Monoclonal Antibody
$200.50 - $1973.00
Specifications
Antigen | PDGF-AA |
---|---|
Clone | 114506 |
Dilution | Neutralization 0.2-1.0 ug/mL |
Applications | Neutralization |
Classification | Monoclonal |
Description
PDGF-AA Monoclonal specifically detects PDGF-AA in Human samples. It is validated for Neutralization.Specifications
PDGF-AA | |
Neutralization 0.2-1.0 ug/mL | |
Monoclonal | |
Mouse | |
Human | |
P04085 | |
5154 | |
E. coli-derived recombinant human PDGF-AA Ser87-Thr211 Accession # P04085 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects human PDGF-AA in direct ELISAs. In direct ELISAs, approximately 20% cross-reactivity with recombinant human (rh) PDGF-AB is observed and no cross-reactivity with rhPDGF-BB, rhPlGF, rhVEGF206, rhVEGF B 186 rhVEGF C, rhVEGF D, or rhCTGF is observed. |
114506 | |
Neutralization | |
Unconjugated | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
PDGF1, PDGF-A, PDGFAA | |
PDGFA | |
IgG1 | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title