Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human PDGF-AA Antibody, R&D Systems™

Mouse Monoclonal Antibody
$188.28 - $698.38
Specifications
| Antigen | PDGF-AA |
|---|---|
| Clone | 114503 |
| Dilution | Western Blot 1 ug/mL |
| Applications | Western Blot |
| Classification | Monoclonal |
Description
PDGF-AA Monoclonal specifically detects PDGF-AA in Human samples. It is validated for Western Blot.Specifications
| PDGF-AA | |
| Western Blot 1 ug/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| PDGF1, PDGF-A, PDGFAA | |
| PDGFA | |
| IgG2b | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| 114503 | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| Human | |
| P04085 | |
| 5154 | |
| E. coli-derived recombinant human PDGF-AA Ser87-Thr211 Accession # P04085 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects human PDGF-AA in direct ELISAs and Western blots. In direct ELISAs, this antibody shows 25% cross-reactivity with recombinant human (rh) PDGF-AB and no cross-reactivity with rhPDGF-BB, rhPlGF, rhVEGF, rhVEGF-C, rhVEGF-D, rhCTGF, or rhLDGF. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title