Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human PDGF-AA Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AB221NA
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AB221NA 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AB221NA Supplier R&D Systems Supplier No. AB221NA
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody

PDGF-AA Polyclonal specifically detects PDGF-AA in Human samples. It is validated for Western Blot, Immunohistochemistry, Neutralization.

Specifications

Antigen PDGF-AA
Applications Western Blot, Immunohistochemistry, Neutralization
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL, Immunohistochemistry 5-15 ug/mL, Neutralization 5-10 ug/mL
Gene Accession No. P04085
Gene Alias PDGF1, PDGF-A, PDGFAA
Gene Symbols PDGFA
Host Species Goat
Immunogen E. coli-derived recombinant human PDGF-AA Ser87-Thr211 Accession # P04085
Purification Method Protein A or G purified
Quantity 1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5154
Test Specificity Detects human PDGF-AA in direct ELISAs and Western blots. Neutralizes the biological activity of human PDGF-AA and will also neutralize the biological activity of recombinant human PDGF-AB, although 2-3 times the amount of IgG is required. It will not neutralize the biological activity of natural human PDGF-AB, porcine PDGF-BB, or rhPDGF-BB.
Reconstitution Reconstitute at 1 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.