Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human PDGF Antibody, R&D Systems™

Rabbit Polyclonal Antibody has been used in 3 publications
Supplier: R&D Systems AB20NA
This item is not returnable.
View return policy
Description
PDGF Polyclonal specifically detects PDGF in Human samples. It is validated for Western Blot, Neutralization.Specifications
| PDGF | |
| Polyclonal | |
| Unconjugated | |
| C-Sis, IBGC5, PDGF1, PDGF-1, PDGF2, PDGF-2, PDGFA, PDGF-A, PDGFAA, PDGFAB, PDGFB, SIS, SSV | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 5154 | |
| Reconstitute at 1 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot, Neutralization | |
| LYOPH | |
| Western Blot 1 ug/mL, Neutralization 5-10 ug/mL | |
| PDGFA | |
| Human platelet-derived PDGF | |
| 1 mg | |
| Primary | |
| Detects human PDGF in direct ELISAs and Western blots. In direct ELISAs, approximately 10% cross-reactivity with recombinant human (rh) PDGF-AA, 60% cross-reactivity with rhPDGF-AB, and nearly 100% cross-reactivity with rhPDGF-BB is observed. | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction