Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human PDGF-B Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB220100
Change view
Click to view available options
Quantity:
100 μg
25 μg
500 μg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB220100 100 μg
MAB220SP 25 μg
MAB220500 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. MAB220100 Supplier R&D Systems Supplier No. MAB220100
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

PDGF-B Monoclonal specifically detects PDGF-B in Human samples. It is validated for Western Blot.

Specifications

Antigen PDGF-B
Applications Western Blot
Classification Monoclonal
Clone 108132
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. P01127
Gene Alias B chain, Becaplermin, C-Sis, FLJ12858, IBGC5, PDGF subunit B, PDGF2, PDGF-2, PDGFB, platelet-derived growth factor 2, Platelet-derived growth factor beta polypeptide, platelet-derived growth factor beta polypeptide (simian sarcoma viral (v-sis)oncogene homolog), platelet-derived growth factor subunit B, platelet-derived growth factor, B chain, Platelet-derived growth factor, beta polypeptide (oncogene SIS), Proto-oncogene c-Sis, SIS, SSV
Gene Symbols PDGFB
Host Species Mouse
Immunogen E. coli-derived recombinant human PDGF-BB Ser82-Thr190 Accession # P01127
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5155
Test Specificity Detects human PDGF-B subunit in direct ELISAs and Western blots. In direct ELISAs, shows approximately 40% cross-reactivity with recombinant human (rh) PDGF-AB and no cross-reactivity with rhCTGF, rhLDGF, rhPDGF-AA, recombinant rat (rr) PDGF-AA, rrPDGF-AB, rrPDGF-BB, rhPDGF-C, rhPDGF-D, rhPlGF, rhVEGF, rhVEGF-C, or rhVEGF-D.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Supernatant
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.