Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human PDGF-BB ELISA Kit

Catalog No. 50246660
Encompass_Preferred
Click to view available options
Quantity:
10 x 96 Tests
96 Tests

Includes

Aluminium pouch(es) with a Microwell Plate coated with antibody to human PDGF-BB
Biotin-Conjugate anti-human PDGF-BB antibody
Streptavidin-HRP
Human PDGF-BB Standard lyophilized, 4ng/mL upon reconstitution
Assay Buffer Concentrate 20x (PBS with 1% Tween 20 and 10% BSA)
Wash Buffer Concentrate 20x (PBS with 1% Tween 20)
Substrate Solution (tetramethyl-benzidine)
Stop Solution (1M Phosphoric acid)
Blue-Dye
Green-Dye
Red-Dye
Adhesive Films

ELISA

The Human Platelet Derived Growth Factor BB (Hu PDGF-BB) ELISA quantitates Hu PDGF-BB in human serum, plasma, buffered solution, or cell culture medium. The assay will exclusively recognize both natural and recombinant Hu PDGF-BB. Principle of the method The Human PDGF-BB solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen. Rigorous validation Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.

The protein encoded by this gene is a member of the platelet-derived growth factor family. The four members of this family are mitogenic factors for cells of mesenchymal origin and are characterized by a motif of eight cysteines. This gene product can exist either as a homodimer (PDGF-BB) or as a heterodimer with the platelet-derived growth factor alpha polypeptide (PDGF-AB), where the dimers are connected by disulfide bonds. Mutations in this gene are associated with meningioma. Reciprocal translocations between chromosomes 22 and 7, at sites where this gene and that for COL1A1 are located, are associated with a particular type of skin tumor called dermatofibrosarcoma protuberans resulting from unregulated expression of growth factor. Two alternatively spliced transcript variants encoding different isoforms have been identified for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P01127
Assay Range 31.3 to 2,000 pg/mL
Assay Sensitivity 4.6 pg/mL
Conjugate HRP
Product Type ELISA
Sample Type Plasma, Serum, Supernatant
Target Species Human
For Use With (Application) ELISA
For Use With (Equipment) Colorimetric Microplate Reader
Gene ID (Entrez) 5155
Gene Symbol IBGC5,PDGF-2,PDGF2,SIS,SSV,c-sis
Interassay CV 8.4%
Intraassay CV 4
Kit Contents Pre-coated 96 well plate, Standard, Sample Diluent, Assay Buffer concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
Label or Dye HRP
Quantity 10 x 96 Tests
Regulatory Status RUO
Sample Volume Plasma 20 μL, Serum 20 μL, Supernatant 20 μL
Storage Requirements 2°C to 8°C
Synonym Platelet derived growth factor BB
Target PDGF-BB
Test Time 1 hrs 20 mins
Total Assay Time 3 hrs 30 mins
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.