Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human PDGF-D Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF1159SP
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF1159SP 25 μg
AF1159 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. AF1159SP Supplier R&D Systems Supplier No. AF1159SP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody has been used in 4 publications

PDGF-D/SCDGFB Polyclonal specifically detects PDGF-D/SCDGFB in Human samples. It is validated for Western Blot, Immunohistochemistry.

Specifications

Antigen PDGF-D/SCDGFB
Applications Western Blot, Immunohistochemistry
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 0.1 ug/mL, Immunohistochemistry 5-15 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. Q9GZP0
Gene Alias IEGFMSTP036, Iris-expressed growth factor, PDGFD, PDGF-D, platelet derived growth factor D, SCDGF-BMGC26867, SCDGFBplatelet-derived growth factor D, spinal cord derived growth factor B, Spinal cord-derived growth factor B, spinal cord-derived growth factor-B
Gene Symbols PDGFD
Host Species Goat
Immunogen E. coli-derived recombinant human PDGF-D Asp253-Arg370 Accession # Q9GZP0
Purification Method Affinity Purified
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 80310
Test Specificity Detects human PDGF-D in direct ELISAs and Western blots. In direct ELISAs and Western blots, less than 1% cross-reactivity with recombinant human (rh) PDGF-AA, rhPDGF-AB and rhPDGF-BB is observed.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.