Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human PDGF-D Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB1159100
Change view
Click to view available options
Quantity:
100 μg
25 μg
500 μg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB1159100 100 μg
MAB1159SP 25 μg
MAB1159500 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. MAB1159100 Supplier R&D Systems Supplier No. MAB1159100
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

PDGF-D/SCDGFB Monoclonal specifically detects PDGF-D/SCDGFB in Human samples. It is validated for Neutralization.

Specifications

Antigen PDGF-D/SCDGFB
Applications Neutralization
Classification Monoclonal
Clone 680018
Concentration LYOPH
Conjugate Unconjugated
Dilution Neutralization 1.5-7.5 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. Q9GZP0.1
Gene Alias IEGFMSTP036, Iris-expressed growth factor, PDGFD, PDGF-D, platelet derived growth factor D, SCDGF-BMGC26867, SCDGFBplatelet-derived growth factor D, spinal cord derived growth factor B, Spinal cord-derived growth factor B, spinal cord-derived growth factor-B
Gene Symbols PDGFD
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human PDGF-DD Ser250-Arg370 Accession # Q9GZP0.1
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 80310
Test Specificity Detects human PDGF-D in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant human (rh) PDGF, rhPDGF-AA, rhPDGF-AB, rhPDGF-BB, rhPDGF-C, or recombinant rat PDGF-AB is observed.
Reconstitution Sterile PBS to a final concentration of 0.5 mg/mL.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Supernatant
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.