Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human PDGF R alpha Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 8 publications
Supplier: R&D Systems MAB1264SP
This item is not returnable.
View return policy
Description
PDGFR alpha Monoclonal specifically detects PDGFR alpha in Human samples. It is validated for Flow Cytometry, Immunoprecipitation.Specifications
PDGFR alpha | |
Monoclonal | |
Unconjugated | |
Supplied as a 0.2 μm filtered solution in PBS. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
PDGFRA | |
Human osteosarcoma cell membrane extracts | |
25 μg | |
Primary | |
Detects human PDGF R alpha . Recognizes the PDGF receptor alpha-subunit; it does not recognize the PDGF receptor beta-subunit. Binds to the PDGF receptor alpha-subunit of primate species (human, monkey, baboon) and dog. Does not recognize rat or mouse receptors and its ability to bind to receptors from other species has not been tested. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC, as supplied. 1 month, 2 to 8 degreesC under sterile conditions after opening. 6 months, -20 to -70 degreesC under sterile conditions after opening. | |
IgG1 |
Flow Cytometry, Immunoprecipitation | |
PRa292 | |
Flow Cytometry 0.25 ug/10^6 cells, Immunoprecipitation 25 ug/mL | |
alpha-type platelet-derived growth factor receptor, CD140 antigen-like family member A, CD140a, CD140a antigen, EC 2.7.10, EC 2.7.10.1, MGC74795, PDGFR alpha, PDGFR2, PDGFRA, PDGFRA/BCR fusion, PDGF-R-alpha, platelet-derived growth factor receptor, alpha polypeptide, rearranged-in-hypereosinophilia-platelet derived growth factor receptor alphafusion protein, RHEPDGFRA | |
Mouse | |
Protein A or G purified from ascites | |
RUO | |
5156 | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction