Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human PDGF R alpha Fluorescein-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems FAB1264F
This item is not returnable.
View return policy
Description
PDGFR alpha Monoclonal specifically detects PDGFR alpha in Human samples. It is validated for Flow Cytometry.Specifications
PDGFR alpha | |
Monoclonal | |
Fluorescein | |
Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide | |
PDGFRA | |
Human osteosarcoma cell membrane extracts | |
100 Tests | |
Primary | |
Detects human PDGF R alpha . Recognizes the PDGF receptor alpha-subunit; it does not recognize the PDGF receptor beta-subunit. Binds to the PDGF receptor alpha-subunit of primate species (human, monkey, baboon) and dog. Does not recognize rat or mouse receptors and its ability to bind to receptors from other species has not been tested. | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. | |
IgG1 |
Flow Cytometry | |
PRa292 | |
Flow Cytometry 10 uL/10^6 cells | |
alpha-type platelet-derived growth factor receptor, CD140 antigen-like family member A, CD140a, CD140a antigen, EC 2.7.10, EC 2.7.10.1, MGC74795, PDGFR alpha, PDGFR2, PDGFRA, PDGFRA/BCR fusion, PDGF-R-alpha, platelet-derived growth factor receptor, alpha polypeptide, rearranged-in-hypereosinophilia-platelet derived growth factor receptor alphafusion protein, RHEPDGFRA | |
Mouse | |
Protein A or G purified from ascites | |
RUO | |
5156 | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction