Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human PDGF R beta Biotinylated Antibody, R&D Systems™

Goat Polyclonal Antibody
Supplier: R&D Systems BAF385
This item is not returnable.
View return policy
Description
PDGF R beta Polyclonal specifically detects PDGF R beta in Human samples. It is validated for Western Blot, Flow Cytometry.Specifications
PDGF R beta | |
Polyclonal | |
Biotin | |
P09619 | |
PDGFRB | |
Mouse myeloma cell line NS0-derived recombinant human PDGF R beta Leu33-Phe530 (Glu241Asp) Accession # P09619 | |
50 μg | |
Primary | |
Detects human PDGF R beta in Western blots. In Western blots, less than 1% cross-reactivity with recombinant human PDGF R alpha is observed. | |
Human | |
IgG |
Western Blot, Flow Cytometry | |
LYOPH | |
Western Blot 0.1 ug/mL, Flow Cytometry 2.5 ug/10^6 cells | |
beta-type platelet-derived growth factor receptor, CD140 antigen-like family member B, CD140b, CD140b antigen, EC 2.7.10, EC 2.7.10.1, JTK12, PDGFR, PDGFR1, PDGFRB, PDGF-R-beta, platelet-derived growth factor receptor, beta polypeptide | |
Goat | |
Affinity Purified | |
RUO | |
5159 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction