Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human PDGF R beta PE-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. FAB1263P
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FAB1263P 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. FAB1263P Supplier R&D Systems Supplier No. FAB1263P
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody has been used in 7 publications

PDGF R beta Monoclonal specifically detects PDGF R beta in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen PDGF R beta
Applications Flow Cytometry
Classification Monoclonal
Clone PR7212
Conjugate R-PE
Dilution Flow Cytometry 10 uL/10^6 cells
Formulation Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide
Gene Alias beta-type platelet-derived growth factor receptor, CD140 antigen-like family member B, CD140b, CD140b antigen, EC 2.7.10, EC 2.7.10.1, JTK12, PDGFR, PDGFR1, PDGFRB, PDGF-R-beta, platelet-derived growth factor receptor, beta polypeptide
Gene Symbols PDGFRB
Host Species Mouse
Immunogen Human skin fibroblast membrane extracts
Purification Method Protein A or G purified from ascites
Quantity 100 Tests
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5159
Test Specificity Detects human PDGF R beta in Western blots. Specificity has been confirmed in binding studies using several different cell lines (1, 2) and by its ability to immunoprecipitate PDGF R beta-subunit complexed with 125 I-PDGF-BB (3). It does not recognize the PDGF R alpha-subunit. Detects the PDGF R beta-subunit of human and primate species (monkey and baboon) but not the rat or mouse receptors. Its ability to bind to receptors from other species has not been tested.
Target Species Human
Content And Storage Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.