Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human PDGFRL Alexa Fluor™ 350-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems FAB1723U100UG
This item is not returnable.
View return policy
Description
PDGFRL Monoclonal antibody specifically detects PDGFRL in Human samples. It is validated for Flow CytometrySpecifications
PDGFRL | |
Monoclonal | |
Alexa Fluor 350 | |
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. | |
PDGFR-like protein, PDGRL, platelet-derived growth factor receptor-like, platelet-derived growth factor receptor-like protein, platelet-derived growth factor-beta-like tumor suppressor, PRLTS, PRLTSPDGF receptor beta-like tumor suppressor | |
Human embryonic kidney cell, HEK293-derived human PDGFRL, Gln22-Ser375, Accession # Q15198 | |
100 μg | |
Primary | |
Human | |
Purified |
Flow Cytometry | |
1008804 | |
Flow Cytometry 0.25-1 ug/106 cells | |
Q15198 | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
5157 | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8°C as supplied. | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction