Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human PDZD2 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems MAB7670
This item is not returnable.
View return policy
Description
PDZD2/PDZK3 Monoclonal specifically detects PDZD2/PDZK3 in Human samples. It is validated for Western Blot, Immunocytochemistry.Specifications
PDZD2/PDZK3 | |
Monoclonal | |
LYOPH | |
Western Blot 2 ug/mL, Immunocytochemistry 8-25 ug/mL | |
O15018 | |
PDZD2 | |
E. coli-derived recombinant human PDZD2 Ser2489-Ser2839 Accession # O15018 | |
100 μg | |
Primary | |
Detects human PDZD2 in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant mouse PDZD2 is observed. | |
Human | |
Purified |
Western Blot, Immunocytochemistry | |
787924 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
Activated in prostate cancer protein, AIPC, AIPCPAPIN, KIAA0300, KIAA0300PDZ domain-containing protein 3, Papin, PDZ domain containing 2, PDZ domain containing 3, PDZ domain-containing protein 2, PDZK3, PIN1 | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
23037 | |
Sterile PBS to a final concentration of 0.5 mg/mL. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction