Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human PEAR1 Alexa Fluor 594-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems FAB4527T100UG
This item is not returnable.
View return policy
Description
PEAR1 Monoclonal specifically detects PEAR1 in Human samples. It is validated for Flow Cytometry.Specifications
PEAR1 | |
Monoclonal | |
0.2 mg/mL | |
Flow Cytometry 0.25-1 ug/10^6 cells | |
Q5VY43 | |
PEAR1 | |
Chinese hamster ovary cell line CHO-derived recombinant human PEAR1 Leu21-Ser754 Accession # Q5VY43 | |
RUO | |
375033 | |
Human | |
IgG2a |
Flow Cytometry | |
492621 | |
Alexa Fluor 594 | |
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
JEDI, platelet endothelial aggregation receptor 1 | |
Mouse | |
100 μg | |
Primary | |
Detects human PEAR1 in direct ELISAs and Western blots. In Western blots, no cross-reactivity with recombinant human Tenascin R or recombinant mouse Tenascin C is observed. | |
Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction