Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human PEAR1 Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 2 publications
Supplier: R&D Systems AF4527SP
This item is not returnable.
View return policy
Description
PEAR1 Polyclonal specifically detects PEAR1 in Human samples. It is validated for Western Blot, Flow Cytometry, CyTOF-ready.Specifications
PEAR1 | |
Polyclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
JEDI, platelet endothelial aggregation receptor 1 | |
Goat | |
Affinity Purified | |
RUO | |
375033 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Western Blot, Flow Cytometry, CyTOF | |
LYOPH | |
Western Blot 0.1 ug/mL, Flow Cytometry 2.5 ug/10^6 cells, CyTOF-ready | |
Q5VY43 | |
PEAR1 | |
Chinese hamster ovary cell line CHO-derived recombinant human PEAR1 Leu21-Ser754 Accession # Q5VY43 | |
25 μg | |
Primary | |
Detects human PEAR1 in direct ELISAs and Western blots. | |
Human | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction