Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human PEDFR/PNPLA2 Alexa Fluor 750-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. FAB5387S100
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FAB5387S100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. FAB5387S100 Supplier R&D Systems Supplier No. FAB5387S100UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

PEDFR/PNPLA2/ATGL Monoclonal specifically detects PEDFR/PNPLA2/ATGL in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen PEDFR/PNPLA2/ATGL
Applications Flow Cytometry
Classification Monoclonal
Clone 494702
Concentration 0.2 mg/mL
Conjugate Alexa Fluor 750
Dilution Flow Cytometry 0.25-1 ug/10^6 cells
Formulation Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide
Gene Accession No. Q96AD5
Gene Alias Adipose triglyceride lipase, ATGL, ATGL1110001C14Rik, Calcium-independent phospholipase A2, Desnutrin, EC 3.1.1.3, FP17548, IPLA2-zeta, patatin-like phospholipase domain containing 2, patatin-like phospholipase domain-containing protein 2, patatin-like phospholipase domain-containing protein 2-like, PEDF R, PEDF-R, Pigment epithelium-derived factor, PNPLA2, Transport-secretion protein 2, transport-secretion protein 2.2, triglyceride hydrolase, TTS2, TTS2.2, TTS-2.2, TTS2DKFZp667M109
Gene Symbols PNPLA2
Host Species Mouse
Immunogen E. coli-derived recombinant human PEDF R/PNPLA2 Val162-Thr332 Accession # Q96AD5
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57104
Test Specificity Detects human PEDF R/PNPLA2 in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant mouse PEDF R is observed.
Target Species Human
Content And Storage Store the unopened product at 2 - 8 degreesC. Do not use past expiration date.
Isotype IgG2b
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.