Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human PEG10 Alexa Fluor 750-conjugated Antibody, R&D Systems™

Rabbit Monoclonal Antibody
Supplier: R&D Systems IC99751S100UG
This item is not returnable.
View return policy
Description
PEG10 Monoclonal specifically detects PEG10 in Human samples. It is validated for Intracellular Staining by Flow Cytometry.Specifications
PEG10 | |
Monoclonal | |
Alexa Fluor 750 | |
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. | |
EDR, embryonal carcinoma differentiation regulated, Embryonal carcinoma differentiation-regulated protein, HB-1, KIAA1051EDR, Mammalian retrotransposon-derived protein 2, Mar2, MART2, MEF3 like 1, MEF3L, MEF3L1, Myelin expression factor 3-like protein 1, paternally expressed 10, Paternally expressed gene 10 protein, retrotransposon gag domain containing 3, Retrotransposon gag domain-containing protein 3, Retrotransposon-derived gag-like polyprotein, retrotransposon-derived protein PEG10, RGAG3, RGAG3MEF3-like protein 1, RTL2, SIRH1, Ty3/Gypsy-like protein | |
Synthetic peptide containing human PEG10, Accession No. Q86TG7 | |
100 μg | |
23089.0 | |
Human | |
Supernatant |
Flow Cytometry | |
981902 | |
Intracellular Staining by Flow Cytometry 0.25-1 μg/106 cells | |
Q86TG7 | |
Rabbit | |
Protein A or G purified from hybridoma culture supernatant | |
Primary | |
Detects human PEG10 in direct ELISAs. | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8°C as supplied. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction