Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human PEG10 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems MAB9975100
This item is not returnable.
View return policy
Description
PEG10 Monoclonal specifically detects PEG10 in Human samples. It is validated for ELISA.Specifications
PEG10 | |
Monoclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
Primary | |
Detects Human PEG10 in direct ELISAs. | |
Human | |
Supernatant |
ELISA | |
981918 | |
ELISA | |
EDR, embryonal carcinoma differentiation regulated, Embryonal carcinoma differentiation-regulated protein, HB-1, KIAA1051EDR, Mammalian retrotransposon-derived protein 2, Mar2, MART2, MEF3 like 1, MEF3L, MEF3L1, Myelin expression factor 3-like protein 1, paternally expressed 10, Paternally expressed gene 10 protein, retrotransposon gag domain containing 3, Retrotransposon gag domain-containing protein 3, Retrotransposon-derived gag-like polyprotein, retrotransposon-derived protein PEG10, RGAG3, RGAG3MEF3-like protein 1, RTL2, SIRH1, Ty3/Gypsy-like protein | |
Synthetic peptide containing human PEG10 | |
100 μg | |
23089.0 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction