Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Pentraxin 2/SAP Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems MAB19481100
This item is not returnable.
View return policy
Description
Pentraxin 2/SAP Monoclonal specifically detects Pentraxin 2/SAP in Human samples. It is validated for ELISA.Specifications
Pentraxin2/SAP | |
Monoclonal | |
Unconjugated | |
Lyophilized from a 0.2μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2μm filtered solution in PBS. | |
9.5S alpha-1-glycoprotein, amyloid P component, serum, APCS, MGC88159, PTX2, PTX2serum amyloid P-component, SAP, SAPpentaxin-related, Serum Amyloid P component | |
Mouse myeloma cell line NS0-derived recombinant human Pentraxin 2/SAP His20-Val233 Accession # P02743 | |
100 μg | |
325 | |
Reconstitute at 0.5mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG1 |
ELISA | |
910115R | |
ELISA | |
P02743 | |
Mouse | |
Protein A or G purified from cell culture supernatant | |
Primary | |
Detects human Pentraxin 2/SAP in direct ELISAs. | |
Human | |
Supernatant |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction