Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Human Pepsinogen A4 DuoSet ELISA
SDP

Catalog No. DY910505 Shop All Novus Products
Change view
Click to view available options
Quantity:
1 Kit (for 5 Plates)
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
DY910505 1 Kit (for 5 Plates)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. DY910505 Supplier R&D Systems™ Supplier No. DY910505

Item Discontinued Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Includes

Capture antibody, detection antibody, recombinant standard, streptavidin conjugated to horseradish-peroxidase (Streptavidin-HRP) and development protocols

For the determination of natural and recombinant Pepsinogen A4 in human. R&D Systems Human Pepsinogen A4 DuoSet ELISA Kit is a solid phase sandwich ELISA suitable for the analysis of cell culture supernate, serum, and plasma samples.

The Human Pepsinogen A4 DuoSet ELISA is a Solid Phase Sandwich ELISA that quantifies human Pepsinogen A4/PGA4.

Specifications

Product Type ELISA Kit
Quantity 1 Kit (for 5 Plates)
Species Human
Binding Target Pepsinogen A4
Assay Range 0.31 to 20 ng/mL
Format 96-well Plates
For Use With (Application) ELISA
Gene ID (Entrez) 643847
Product Line R&D Systems
Sample Type Cell Culture Supernatant, Serum, Plasma
Content And Storage Store the unopened product at 2°C to 8°C. Do not use past expiration date.
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.