Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Pepsinogen A5/PGA5 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF8457100
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF8457100 100 μg
AF8457SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. AF8457100 Supplier R&D Systems Supplier No. AF8457100
Add to Cart
Add to Cart
This item is not returnable. View return policy

Sheep Antibody

PGA5 specifically detects PGA5 in Human samples. It is validated for Western Blot, Simple Western.

Specifications

Antigen PGA5
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 0.1 μg;/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS.
Gene Accession No. P0DJD7
Gene Alias EC 3.4.23, EC 3.4.23.1, pepsin A, pepsinogen 5, group I (pepsinogen A), pepsinogen A5, PGA3, PGA4, PGA5
Host Species Sheep
Immunogen Mouse myeloma cell line NS0-derived recombinant human Pepsinogen A5/PGA5, Met1-Ala388, Accession No. P0DJD7
Purification Method Antigen Affinity-purified
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) 5222.0
Test Specificity Detects human Pepsinogen A5/PGA5 in direct ELISAs and Western blots.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution.
Form Lyophilized
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.