Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Peptidase Inhibitor 16/PI16 Antibody, R&D Systems™

Goat Polyclonal Antibody
Supplier: R&D Systems AF4980SP
This item is not returnable.
View return policy
Description
Peptidase Inhibitor 16/PI16 Polyclonal specifically detects Peptidase Inhibitor 16/PI16 in Human samples. It is validated for Western Blot.Specifications
Peptidase Inhibitor 16/PI16 | |
Polyclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
CRISP9, CRISP-9, Cysteine-rich secretory protein 9, dJ90K10.5, DKFZp586B1817, MGC45378, microseminoprotein, beta-binding protein, MSMBBP, peptidase inhibitor 16, PI16, PI-16, protease inhibitor 16, PSP94-binding protein, PSPBP | |
Goat | |
Antigen Affinity-purified | |
RUO | |
221476 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Western Blot | |
LYOPH | |
Western Blot 0.1 ug/mL | |
AAH35634 | |
PI16 | |
Mouse myeloma cell line NS0-derived recombinant human Peptidase Inhibitor 16/PI16 Leu28-Leu437 Accession # AAH35634 | |
25 μg | |
Primary | |
Detects human Peptidase Inhibitor 16/PI16 in direct ELISAs and Western blots. | |
Human | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction