Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Peptide YY Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB9456100
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB9456100 100 μg
MAB9456SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. MAB9456100 Supplier R&D Systems Supplier No. MAB9456100
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Monoclonal Antibody

Peptide YY Monoclonal specifically detects Peptide YY in Human samples. It is validated for Immunohistochemistry.

Specifications

Antigen Peptide YY
Applications Immunohistochemistry
Classification Monoclonal
Clone 2163A
Conjugate Unconjugated
Dilution Immunohistochemistry 3-10 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS.
Gene Accession No. P10082
Gene Alias Peptide tyrosine tyrosine, peptide YY, Peptide-YY, PYY, PYY1, PYY-I
Host Species Rabbit
Immunogen Human Peptide YY synthetic peptide Accession # P10082
Purification Method Protein A or G purified from cell culture supernatant
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5697
Test Specificity Detects human Peptide YY.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.