Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human PER1 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF4565
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF4565 100 μg
AF4565SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. AF4565 Supplier R&D Systems Supplier No. AF4565
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody

PER1 Polyclonal specifically detects PER1 in Human samples. It is validated for Immunohistochemistry.

Specifications

Antigen PER1
Applications Immunohistochemistry
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Immunohistochemistry 5-15 ug/mL
Gene Accession No. O15534
Gene Alias Circadian clock protein PERIOD 1, Circadian pacemaker protein Rigui, hPER1, KIAA0482, PERhPER, period (Drosophila) homolog 1, period circadian protein homolog 1, period homolog 1 (Drosophila), Period, drosophila, homolog of, PERIOD1, RIGUI, RIGUIMGC88021
Gene Symbols PER1
Host Species Goat
Immunogen E. coli-derived recombinant human PER1 Ser159-Thr365 Accession # O15534
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5187
Test Specificity Detects human PER1 in direct ELISAs.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.