Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Perforin Antibody, R&D Systems™

Mouse Monoclonal Antibody
$241.00 - $549.50
Specifications
Antigen | Perforin |
---|---|
Clone | 1001103 |
Dilution | Western Blot 2 μg/mL, Immunohistochemistry 5-25 μg/mL |
Applications | Western Blot, Immunohistochemistry |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB8011SP
![]() |
R&D Systems
MAB8011SP |
25 μg |
Each for $241.00
|
|
|||||
MAB8011100
![]() |
R&D Systems
MAB8011100 |
100 μg |
Each for $549.50
|
|
|||||
Description
Perforin Monoclonal specifically detects Perforin in Human samples. It is validated for Western Blot, Immunohistochemistry.Specifications
Perforin | |
Western Blot 2 μg/mL, Immunohistochemistry 5-25 μg/mL | |
Monoclonal | |
Mouse | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
Cytolysin, FLH2, HPLH2, HPLH2lymphocyte pore forming protein, Lymphocyte pore-forming protein, MGC65093, P1, P1PFN1, perforin 1 (pore forming protein), perforin-1, PFP, PFPcytolysin, PRF1 | |
Synthetic peptide containing human Perforin, Pro22-Trp555, Accession No. P14222 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects human Perforin in direct ELISAs. |
1001103 | |
Western Blot, Immunohistochemistry | |
Unconjugated | |
Human | |
P14222 | |
5551.0 | |
IgG2a | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title