Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Perilipin-3 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB7664SP
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB7664SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. MAB7664SP Supplier R&D Systems Supplier No. MAB7664SP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

Perilipin-3/TIP47 Monoclonal specifically detects Perilipin-3/TIP47 in Human samples. It is validated for Western Blot.

Specifications

Antigen Perilipin-3/TIP47
Applications Western Blot
Classification Monoclonal
Clone 779119
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 0.5 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. O60664
Gene Alias Cargo selection protein TIP47, M6PRBP1, M6PRBP1MGC2012, Mannose-6-phosphate receptor-binding protein 1, perilipin 3,47 kDa MPR-binding protein, perilipin-3, Perilipin3, Placental protein 17,47 kDa mannose 6-phosphate receptor-binding protein, PLIN3, PP17, PP17MGC11117, tail-interacting protein, 47 kD, TIP47, TIP47mannose-6-phosphate receptor binding protein 1
Gene Symbols PLIN3
Host Species Mouse
Immunogen E. coli-derived recombinant human Perilipin-3 Met1-Lys434 Accession # O60664
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10226
Test Specificity Detects human Perilipin-3 in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant human (rh) Perilipin-2 or rhPerilipin-5 is observed.
Reconstitution Sterile PBS to a final concentration of 0.5 mg/mL.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2b
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.