Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Periostin/OSF-2 Antibody, R&D Systems™

Mouse Monoclonal Antibody
$241.00 - $549.50
Specifications
Antigen | Periostin/OSF-2 |
---|---|
Clone | 613618R |
Dilution | ELISA |
Applications | ELISA |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB35483SP
![]() |
R&D Systems
MAB35483SP |
25 μg |
Each for $241.00
|
|
|||||
MAB35483100
![]() |
R&D Systems
MAB35483100 |
100 μg |
Each for $549.50
|
|
|||||
Description
Periostin/OSF-2 Monoclonal specifically detects Periostin/OSF-2 in Human samples. It is validated for ELISA.Specifications
Periostin/OSF-2 | |
ELISA | |
Monoclonal | |
Supernatant | |
Human | |
Q15063 | |
10631 | |
IgG1 | |
Protein A or G purified from cell culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
613618R | |
ELISA | |
Unconjugated | |
Mouse | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
Fasciclin I-like, MGC119510, MGC119511, OSF2, OSF-2, OSF-2osteoblast specific factor 2 (fasciclin I-like), OSF2periodontal ligament-specific periostin, Osteoblast-specific factor 2, PDLPOSTN, periostin, periostin isoform thy2, periostin isoform thy4, periostin isoform thy6, periostin isoform thy8, periostin, osteoblast specific factor, PNRP11-412K4.1, POSTN, TRIF52 | |
Mouse myeloma cell line NS0-derived recombinant human Periostin/OSF-2 Met1-Gln836 Accession # Q15063 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects human Periostin/OSF-2 in direct ELISAs. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title