Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Periostin/OSF-2 Antibody, R&D Systems™

Goat Polyclonal Antibody
Supplier: R&D Systems AF3548
This item is not returnable.
View return policy
Description
Periostin/OSF-2 Polyclonal specifically detects Periostin/OSF-2 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation.Specifications
Periostin/OSF-2 | |
Polyclonal | |
Unconjugated | |
Q15063 | |
POSTN | |
Mouse myeloma cell line NS0-derived recombinant human Periostin Asn22-Gln836 Accession # Q15063 | |
100 μg | |
Primary | |
Detects human Periostin/OSF-2 in direct ELISAs and Western blots. In direct ELISAs, approximately 45% cross-reactivity with recombinant mouse Periostin is observed. | |
Human | |
IgG |
Western Blot, Immunohistochemistry, Immunoprecipitation | |
LYOPH | |
Western Blot 1 ug/mL, Immunohistochemistry 5-15 ug/mL, Immunoprecipitation 25 ug/mL | |
Fasciclin I-like, MGC119510, MGC119511, OSF2, OSF-2, OSF-2osteoblast specific factor 2 (fasciclin I-like), OSF2periodontal ligament-specific periostin, Osteoblast-specific factor 2, PDLPOSTN, periostin, periostin isoform thy2, periostin isoform thy4, periostin isoform thy6, periostin isoform thy8, periostin, osteoblast specific factor, PNRP11-412K4.1, POSTN, TRIF52 | |
Goat | |
Affinity Purified | |
RUO | |
10631 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction