Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human PERK Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 3 publications
Supplier: R&D Systems AF3999SP
This item is not returnable.
View return policy
Description
PERK Polyclonal specifically detects PERK in Human samples. It is validated for Western Blot, Knockout Validated.Specifications
PERK | |
Polyclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
DKFZp781H1925, EC 2.7.11.1, EIF2AK3, eukaryotic translation initiation factor 2 alpha kinase 3, eukaryotic translation initiation factor 2-alpha kinase 3, hsPEK, PEK, PEKPERKPancreatic eIF2-alpha kinase, PRKR-like endoplasmic reticulum kinase, WRS | |
Goat | |
Affinity Purified | |
RUO | |
9451 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Western Blot, KnockDown | |
LYOPH | |
Western Blot 1 ug/mL, Knockout Validated | |
Q9NZJ5 | |
EIF2AK3 | |
E. coli-derived recombinant human PERK Ala29-Gln230 Accession # Q9NZJ5 | |
25 μg | |
Primary | |
Detects human PERK in Western blots. | |
Human | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction