Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human PGLYRP1/PGRP-S Biotinylated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. BAF2590
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BAF2590 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. BAF2590 Supplier R&D Systems Supplier No. BAF2590
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody

PGLYRP1/PGRP-S Polyclonal specifically detects PGLYRP1/PGRP-S in Human samples. It is validated for Western Blot, ELISA Detection (Matched Antibody Pair).

Specifications

Antigen PGLYRP1/PGRP-S
Applications Western Blot, ELISA
Classification Polyclonal
Concentration LYOPH
Conjugate Biotin
Dilution Western Blot 0.1 ug/mL, ELISA Detection (Matched Antibody Pair) 0.1-0.4 ug/mL
Gene Accession No. O75594
Gene Alias peptidoglycan recognition protein 1, Peptidoglycan recognition protein short, PGLYRP, PGRPpeptidoglycan recognition protein, PGRPS, PGRP-S, PGRP-SMGC126894, PGRPSMGC126896, TAG7, TNF superfamily, member 3 (LTB)-like (peptidoglycan recognition protein), TNFSF3L
Gene Symbols PGLYRP1
Host Species Goat
Immunogen Mouse myeloma cell line NS0-derived recombinant human PGLYRP1/PGRP-S Gln22-Pro196 Accession # O75594
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8993
Test Specificity Detects human PGLYRP1/PGRP-S in ELISAs and Western blots. In sandwich Immunoassays, less than 0.05% cross-reactivity with recombinant human PGRP-1 beta and recombinant mouse PGRP-S is observed.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.