Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human PGLYRP4/PGRP-I beta Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF3018SP
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF3018SP 25 μg
AF3018 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. AF3018SP Supplier R&D Systems Supplier No. AF3018SP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody

PGLYRP4/PGRP-I beta Polyclonal specifically detects PGLYRP4/PGRP-I beta in Human samples. It is validated for Western Blot.

Specifications

Antigen PGLYRP4/PGRP-I beta
Applications Western Blot
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 0.1 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. Q96LB8
Gene Alias peptidoglycan recognition protein 4, peptidoglycan recognition protein I beta, Pglyrp-1 beta, PGRPI beta, PGRP-I beta, PGRP-I-beta
Gene Symbols PGLYRP4
Host Species Goat
Immunogen Mouse myeloma cell line NS0-derived recombinant human PGLYRP4/PGRP-I beta Asp18-Lys372 Accession # Q96LB8
Purification Method Affinity Purified
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57115
Test Specificity Detects human PGLYRP4/PGRP-I beta in direct ELISAs and Western blots. In direct ELISAs and Western blots, approximately 50% cross-reactivity with recombinant mouse PGRP-I beta is observed and 10% cross-reactivity with recombinant human PGRP-S is observed.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.