Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human PGLYRP4/PGRP-I beta Biotinylated Antibody, R&D Systems™

Goat Polyclonal Antibody
Supplier: R&D Systems BAF3018
This item is not returnable.
View return policy
Description
PGLYRP4/PGRP-I beta Polyclonal specifically detects PGLYRP4/PGRP-I beta in Human samples. It is validated for Western Blot.Specifications
PGLYRP4/PGRP-I beta | |
Polyclonal | |
Biotin | |
AAI07158 | |
PGLYRP4 | |
Mouse myeloma cell line NS0-derived recombinant human PGLYRP4/PGRP-I beta Asp18-Lys372 Accession # AAI07158 | |
50 μg | |
Primary | |
Detects human PGLYRP4/PGRP-I beta in Western blots. In Western blots, approximately 50% cross-reactivity with recombinant mouse PGRP-I beta is observed and 10% cross-reactivity with recombinant human PGRP-S is observed. | |
Human | |
IgG |
Western Blot | |
LYOPH | |
Western Blot 0.1 ug/mL | |
peptidoglycan recognition protein 4, peptidoglycan recognition protein I beta, Pglyrp-1 beta, PGRPI beta, PGRP-I beta, PGRP-I-beta | |
Goat | |
Affinity Purified | |
RUO | |
57115 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction