Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human PHF20 (aa 537-638) Control Fragment Recombinant Protein

Catalog No. RP95252
Encompass_Preferred
Click to view available options
Quantity:
100 μL

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (90%), Rat (90%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-56211 (PA5-56211. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

Methyllysine-binding protein, component of the MOF histone acetyltransferase protein complex. Not required for maintaining the global histone H4 'Lys-16' acetylation (H4K16ac) levels or locus specific histone acetylation, but instead works downstream in transcriptional regulation of MOF target genes. As part of the NSL complex it may be involved in acetylation of nucleosomal histone H4 on several lysine residues. Contributes to methyllysine-dependent p53/TP53 stabilization and up-regulation after DNA damage.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q9BVI0
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 51230
Name Human PHF20 (aa 537-638) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias 6820402O20Rik; C20orf104; GLEA2; Glioma-expressed antigen 2; HCA58; Hepatocellular carcinoma-associated antigen 58; hepatocellular carcinoma-associated antigen 58 homolog; Novel zinc finger protein; NZF; PHD finger 20-like protein; PHD finger protein 20; PHD finger protein 20-like protein; Phf20; TDRD20A; transcription factor TZP; tudor domain containing 20 A; TZP
Common Name PHF20
Gene Symbol PHF20
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence VRVKPKKKKKKKKKTKPECPCSEEISDTSQEPSPPKAFAVTRCGSSHKPGVHMSPQLHGPESGHHKGKVKALEEDNLSESSSESFLWSDDEYGQDVDVTTNP
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.