Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Phospho-53BP1 (S25) Antibody, R&D Systems™

Rabbit Polyclonal Antibody
Supplier: R&D Systems AF3405SP
This item is not returnable.
View return policy
Description
53BP1 Polyclonal specifically detects 53BP1 in Human samples. It is validated for Western Blot.Specifications
53BP1 (p ser25) | |
Polyclonal | |
Western Blot 1 ug/mL | |
53BP1, p202, p53-binding protein 1, p53bp1, TDRD30, TP53-Binding Protein 1, TP53BP1, tumor protein 53-binding protein, 1, tumor protein p53 binding protein 1, tumor protein p53-binding protein, 1, tumor suppressor p53-binding protein 1 | |
Rabbit | |
Affinity Purified | |
RUO | |
7158 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Western Blot | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
TP53BP1 | |
Phosphopeptide containing human 53BP1 S25 site | |
25 μg | |
Primary | |
Detects human 53BP1 when phosphorylated at S25. | |
Human | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction