Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Human Phospho-AMPK alpha 1 (T183) DuoSet IC ELISA

Catalog No. DYC3528E
Change view
Click to view available options
Packaging Quantity:
15 Plates
2 Plates
5 Plates
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Packaging Quantity
DYC3528E 15 Plates
DYC35282 2 Plates
DYC35285 5 Plates
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. DYC3528E Supplier R&D Systems™ Supplier No. DYC3528E
This item is not returnable. View return policy

contains the basic components required for the development of sandwich ELISAs to measure in cell lysates

  • Optimized capture and detection antibody pairings with recommended concentrations save lengthy development time
  • Development protocols are provided to guide further assay optimization
  • Assay can be customized to your specific needs

Specifications

Product Type ELISA Development Kit for Phosphorylated Intracellular Proteins
Quantity 1 Kit
Species Human
Cross Reactivity <50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules
Gene ID (Entrez) 5562
Packaging Quantity 15 Plates
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.