Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Phospho-BTK (Y223) Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems MAB7205
This item is not returnable.
View return policy
Description
BTK Monoclonal specifically detects BTK in Human samples. It is validated for Western Blot, Immunocytochemistry.Specifications
BTK (p tyr223) | |
Monoclonal | |
LYOPH | |
Western Blot 1 ug/mL, Immunocytochemistry 8-25 ug/mL | |
Agammaglobulinaemia tyrosine kinase, AGMX1, AGMX1MGC126262, AT, ATKIMD1, B-cell progenitor kinase, BPK, Bruton agammaglobulinemia tyrosine kinase, Bruton tyrosine kinase, dominant-negative kinase-deficient Brutons tyrosine kinase, EC 2.7.10, EC 2.7.10.2, IMD1, PSCTK1, tyrosine-protein kinase BTK, XLA, XLAMGC126261 | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
695 | |
Sterile PBS to a final concentration of 0.5 mg/mL. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG2b |
Western Blot, Immunocytochemistry | |
720101 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
BTK | |
Phosphopeptide containing the human BTK Y223 site | |
100 μg | |
Primary | |
Detects human Phospho-BTK (Y223) in direct ELISAs and Western blots. | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction