Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Phospho-CREB (S133) Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems MAB6906SP
This item is not returnable.
View return policy
Description
CREB Monoclonal specifically detects CREB in Human samples. It is validated for Western Blot, Immunocytochemistry.Specifications
CREB (p ser133) | |
Monoclonal | |
LYOPH | |
Western Blot 0.5 ug/mL, Immunocytochemistry 8-25 ug/mL | |
active transcription factor CREB, cAMP responsive element binding protein 1, cAMP-response element-binding protein-1, cAMP-responsive element-binding protein 1, CREB, CREB1, CREB-1, cyclic AMP-responsive element-binding protein 1, MGC9284, transactivator protein | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
1385 | |
Sterile PBS to a final concentration of 0.5 mg/mL. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG2a |
Western Blot, Immunocytochemistry | |
702710 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
CREB1 | |
Phosphopeptide containing the human CREB S133 site | |
25 μg | |
Primary | |
Detects human CREB when phosphorylated at S133. | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction