Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Human Phospho-DDR1 DuoSet IC ELISA
SDP

Catalog No. DYC58595 Shop All R&D Systems Products
Change view
Click to view available options
Packaging Quantity:
15 Plates
2 Plates
5 Plates
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Packaging Quantity
DYC58595 5 Plates
DYC58592 2 Plates
DYC5859E 15 Plates
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. DYC58595 Supplier R&D Systems™ Supplier No. DYC58595
Add to Cart
Add to Cart
This item is not returnable. View return policy

contains the basic components required for the development of sandwich ELISAs to measure in cell lysates

The Human Phospho-DDR1 DuoSet IC ELISA is a Solid Phase Sandwich ELISA that quantifies phosphorylated human DDR1.

Specifications

Product Type ELISA Development Kit for Phosphorylated Intracellular Proteins
Quantity 1 Kit
Species Human
Conjugate Biotin
For Use With (Application) ELISA
Gene ID (Entrez) 780
Packaging Quantity 5 Plates
Content And Storage Store the unopened product at 2°C to 8°C. Do not use past expiration date.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.