Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Phospho-eNOS (S1177) Antibody, R&D Systems™

Rabbit Monoclonal Antibody
$212.05 - $556.27
Specifications
| Antigen | eNOS |
|---|---|
| Clone | 1014F |
| Dilution | Western Blot 0.5 ug/mL, Simple Western 10 ug/mL |
| Applications | Western Blot |
| Classification | Monoclonal |
Description
eNOS Monoclonal specifically detects eNOS in Human samples. It is validated for Western Blot, Simple Western.Specifications
| eNOS | |
| Western Blot 0.5 ug/mL, Simple Western 10 ug/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with Sodium Azide | |
| Constitutive NOS, EC 1.14.13.39, ECNOS, endothelial, Endothelial NOS, eNOScNOS, nitric oxide synthase 3 (endothelial cell), NOS type III, NOS3, NOSIII | |
| NOS3 | |
| IgG | |
| Protein A or G purified from cell culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| 1014F | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human | |
| P29474 | |
| 4846 | |
| Phosphopeptide containing the human eNOS S1177 site Accession # P29474 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS | |
| Detects human eNOS when phosphorylated at S1177 in direct ELISAs and Western blots. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title