Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Phospho-ER alpha/NR3A1 (S118) Antibody, R&D Systems™

Rabbit Polyclonal Antibody
Supplier: R&D Systems AF5954
This item is not returnable.
View return policy
Description
ER alpha/NR3A1 Polyclonal specifically detects ER alpha/NR3A1 in Human samples. It is validated for Western Blot.Specifications
ER alpha/NR3A1 (p ser118) | |
Polyclonal | |
Unconjugated | |
P03372 | |
ESR1 | |
Phosphopeptide containing the human ER alpha/NR3A1 S118 site Accession # P03372 | |
100 μg | |
Primary | |
Detects human Phospho-ER alpha/NR3A1 (S118) in Western blots. | |
Human | |
IgG |
Western Blot | |
LYOPH | |
Western Blot 1 ug/mL | |
ER, ER alpha, Era, ER-alpha, ESR1, ESRESRA, Estradiol receptor, estrogen receptor, estrogen receptor 1, estrogen receptor alpha, estrogen receptor alpha delta 3*4,56,7*/819-2 isoform, estrogen receptor alpha delta 4 +49 isoform, estrogen receptor alpha delta 4*5,6,7*/654 isoform, NR3A1, NR3A1DKFZp686N23123, Nuclear receptor subfamily 3 group A member 1 | |
Rabbit | |
Affinity Purified | |
RUO | |
2099 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction