Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Human Phospho-ErbB2/Her2 DuoSet IC ELISA
SDP

Catalog No. DYC1768E Shop All Novus Biologicals Products
Change view
Click to view available options
Packaging Quantity:
15 Plates
2 Plates
5 Plates
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Packaging Quantity
DYC1768E 15 Plates
DYC17685 5 Plates
DYC17682 2 Plates
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. DYC1768E Supplier R&D Systems™ Supplier No. DYC1768E

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Human Phospho-ErbB2/Her2 DuoSet IC ELISA from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.

The Human Phospho-ErbB2/Her2 DuoSet IC ELISA is a Solid Phase Sandwich ELISA that quantifies phosphorylated human ErbB2/Her2.

Specifications

Product Type ELISA Development Kit for Phosphorylated Intracellular Proteins
Quantity 1 Kit
Species Human
Conjugate Biotin
For Use With (Application) ELISA
Gene ID (Entrez) 2064
Product Line R&D Systems
Packaging Quantity 15 Plates
Content And Storage Store the unopened product at 2°C to 8°C. Do not use past expiration date.
Research Area Signaling
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.