Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Phospho-ErbB2/Her2 (Y1248) Antibody, R&D Systems™

Rabbit Polyclonal Antibody has been used in 2 publications
Supplier: R&D Systems AF1768
This item is not returnable.
View return policy
Description
ErbB2/Her2 Polyclonal specifically detects ErbB2/Her2 in Human samples. It is validated for Western Blot, Simple Western, Immunohistochemistry.Specifications
ErbB2/Her2 (p Tyr1248) | |
Polyclonal | |
Unconjugated | |
CD340, CD340 antigen, c-erb B2/neu protein, EC 2.7.10, EGFR2, HER2, HER-2, HER2EC 2.7.10.1, herstatin, Metastatic lymph node gene 19 protein, MLN 19, MLN19, Neu Oncogene, NEUHER-2/neu, neuroblastoma/glioblastoma derived oncogene homolog, NGL, NGLTKR1, p185erbB2, Proto-oncogene c-ErbB-2, Proto-oncogene Neu, receptor tyrosine-protein kinase erbB-2, TKR1, Tyrosine kinase-type cell surface receptor HER2, v-erb-b2 avian erythroblastic leukemia viral oncogene homolog 2(neuro/glioblastoma derived oncogene homolog), v-erb-b2 erythroblastic leukemia viral oncogene homolog 2, neuro/glioblastomaderived oncogene homolog (avian) | |
Rabbit | |
Affinity Purified | |
RUO | |
2064 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Western Blot, Immunohistochemistry | |
LYOPH | |
Western Blot 0.25 ug/mL, Simple Western 5 ug/mL, Immunohistochemistry 0.3-15 ug/mL | |
ERBB2 | |
Phosphopeptide containing human ErbB2 Y1248 site | |
50 μg | |
Primary | |
Detects human ErbB2 when phosphorylated at Y1248. | |
Human | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction