Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Phospho-Erythropoietin R (Y426) Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems MAB6926SP
This item is not returnable.
View return policy
Description
Erythropoietin R Monoclonal specifically detects Erythropoietin R in Human samples. It is validated for Western Blot, Simple Western.Specifications
Erythropoietin R (p Tyr426) | |
Monoclonal | |
LYOPH | |
Western Blot 1 ug/mL, Simple Western 10 ug/mL | |
P19235 | |
EPOR | |
Phosphopeptide containing the human Erythropoietin R Y426 site Accession # P19235 | |
25 μg | |
Primary | |
Detects human Phospho-Erythropoietin R (Y426) in direct ELISAs and Western blots. | |
Human | |
Purified |
Western Blot | |
690710 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
EpoR, EPO-R, erythropoietin receptor, MGC138358 | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
2057 | |
Sterile PBS to a final concentration of 0.5 mg/mL. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction