Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Phospho-FADD (S194) Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems MAB7047SP
This item is not returnable.
View return policy
Description
FADD Monoclonal specifically detects FADD in Human samples. It is validated for Western Blot, Immunocytochemistry.Specifications
FADD (p ser194) | |
Monoclonal | |
LYOPH | |
Western Blot 1 ug/mL, Immunocytochemistry 8-25 ug/mL | |
Fas (TNFRSF6)-associated via death domain, FAS-associated death domain protein, FAS-associating death domain-containing protein, GIG3, Growth-inhibiting gene 3 protein, Mediator of receptor induced toxicity, mediator of receptor-induced toxicity, MGC8528, MORT1, MORT1Fas-associating protein with death domain, protein FADD | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
8772 | |
Sterile PBS to a final concentration of 0.5 mg/mL. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG2a |
Western Blot, Immunocytochemistry | |
709736 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
FADD | |
Phosphopeptide containing the human FADD S194 site | |
25 μg | |
Primary | |
Detects human FADD when phosphorylated on Ser194. | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction